5-Methoxy-4-methylbenzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

118370-1G 118370-500MG 118370-5G
77464-174EA 562.38 USD
77464-174 77476-966 77487-048
5-Methoxy-4-methylbenzo[d][1,3]dioxole ≥95%
5-Methoxy-4-methylbenzo[d][1,3]dioxole
Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD23709618
CAS Number: 607351-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR