1-(4-Bromo-1-fluoronaphthalen-2-yl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

118300-1G 118300-500MG 118300-5G 118300-25G
77464-166EA 415.09 USD
77464-166 77476-958 77487-040 77497-884
1-(4-Bromo-1-fluoronaphthalen-2-yl)-2-methylpropan-1-ol ≥95%
1-(4-Bromo-1-fluoronaphthalen-2-yl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₁₄BrFO
MW: 297.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784045
CAS Number: 2918893-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR