1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

118282-1G 118282-500MG 118282-5G 118282-25G
77464-162EA 415.09 USD
77464-162 77476-954 77487-036 77497-882
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-methylpropan-1-ol ≥95%
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₆H₁₆BrFO
MW: 323.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784028
CAS Number: 2918912-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR