1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

118265-1G 118265-500MG 118265-5G 118265-25G
77464-158EA 415.09 USD
77464-158 77476-950 77487-030 77497-876
1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methylpropan-1-ol ≥95%
1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFOS
MW: 293.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784014
CAS Number: 2918843-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR