(2,6-Difluoro-3-iodophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118244-1G 118244-500MG 118244-5G 118244-25G
77464-154EA 415.09 USD
77464-154 77476-946 77487-026 77497-874
(2,6-Difluoro-3-iodophenyl)(phenyl)methanol ≥95%
(2,6-Difluoro-3-iodophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₉F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34783995
CAS Number: 2918863-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR