(2,6-Difluoro-4-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118220-1G 118220-500MG 118220-5G 118220-25G
77464-150EA 415.09 USD
77464-150 77476-942 77487-022 77497-870
(2,6-Difluoro-4-methoxyphenyl)(phenyl)methanol ≥95%
(2,6-Difluoro-4-methoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16356303
CAS Number: 1274691-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR