(2-Fluoro-5-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118206-1G 118206-500MG 118206-5G
77464-146EA 447.02 USD
77464-146 77476-938 77487-018
(2-Fluoro-5-methoxyphenyl)(phenyl)methanol ≥95%
(2-Fluoro-5-methoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783959
CAS Number: 1204772-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR