(5-Bromo-2-fluoro-3-(methylthio)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118202-1G 118202-500MG 118202-5G 118202-25G
77464-142EA 415.09 USD
77464-142 77476-934 77487-014 77497-864
(5-Bromo-2-fluoro-3-(methylthio)phenyl)(phenyl)methanol ≥95%
(5-Bromo-2-fluoro-3-(methylthio)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₂BrFOS
MW: 327.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783957
CAS Number: 2918843-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR