(3-Chloropyridin-4-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118197-1G 118197-500MG 118197-5G 118197-25G
77464-138EA 307.97 USD
77464-138 77476-930 77487-010 77497-860
(3-Chloropyridin-4-yl)(phenyl)methanol ≥95%
(3-Chloropyridin-4-yl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₂H₁₀ClNO
MW: 219.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998247
CAS Number: 77332-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR