(2-Bromo-3-fluoropyridin-4-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118175-1G 118175-500MG 118175-5G 118175-25G
77464-134EA 415.09 USD
77464-134 77476-926 77487-006 77497-856
(2-Bromo-3-fluoropyridin-4-yl)(phenyl)methanol ≥95%
(2-Bromo-3-fluoropyridin-4-yl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₂H₉BrFNO
MW: 282.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783931
CAS Number: 2918913-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR