(2-Bromo-6-chlorophenyl)(phenyl)methanol ≥96%

Supplier: AOBChem USA

Warning

118165-1G 118165-500MG 118165-5G
77464-130EA 447.02 USD
77464-130 77476-922 77487-002
(2-Bromo-6-chlorophenyl)(phenyl)methanol ≥96%
(2-Bromo-6-chlorophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₀BrClO
MW: 297.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783921
CAS Number: 134456-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR