1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone ≥96%

Supplier: AOBChem USA

Warning

117742-1G 117742-500MG 117742-5G 117742-25G
77464-126EA 415.09 USD
77464-126 77476-918 77486-998 77497-846
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone ≥96%
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₅BrClF₃O₂
MW: 317.4870 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823513
CAS Number: 2918953-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR