1-(3-Bromo-2,6-difluorophenyl)-2-chloro-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

117735-1G 117735-500MG 117735-5G 117735-25G
77464-122EA 397.58 USD
77464-122 77476-914 77486-994 77497-844
1-(3-Bromo-2,6-difluorophenyl)-2-chloro-2,2-difluoroethanone ≥97%
1-(3-Bromo-2,6-difluorophenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂BrClF₄O
MW: 305.4514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29490639
CAS Number: 1698813-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR