1-(2,6-dibromophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

117463-1G 117463-500MG 117463-5G
77464-118EA 447.02 USD
77464-118 77476-910 77486-990
1-(2,6-dibromophenyl)-2-methylpropan-1-ol ≥97%
1-(2,6-dibromophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34783496
CAS Number: 2761573-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR