1-(2-Bromo-5-fluoropyridin-4-yl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117454-1G 117454-500MG 117454-5G 117454-25G
77464-114EA 415.09 USD
77464-114 77476-906 77486-986 77497-838
1-(2-Bromo-5-fluoropyridin-4-yl)-2-methylpropan-1-ol ≥95%
1-(2-Bromo-5-fluoropyridin-4-yl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₉H₁₁BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34783488
CAS Number: 2918892-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR