1-(6-Bromo-2,3,4-trifluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117444-1G 117444-500MG 117444-5G
77464-110EA 546.93 USD
77464-110 77476-902 77486-982
1-(6-Bromo-2,3,4-trifluorophenyl)-2-methylpropan-1-ol ≥95%
1-(6-Bromo-2,3,4-trifluorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₀BrF₃O
MW: 283.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783480
CAS Number: 2024042-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR