Ethyl 2-(furan-3-yl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

117293-1G 117293-500MG 117293-5G 117293-25G
77464-102EA 397.58 USD
77464-102 77476-894 77486-974 77497-830
Ethyl 2-(furan-3-yl)-2-oxoacetate ≥97%
Ethyl 2-(furan-3-yl)-2-oxoacetate

Precaución: For research use only

Formula: C₈H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD16618263
CAS Number: 1379271-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR