2',6'-dimethoxy-[1,1'-biphenyl]-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

117227-1G 117227-500MG 117227-5G 117227-25G
77464-098EA 279.13 USD
77464-098 77476-890 77486-970 77497-826
2',6'-dimethoxy-[1,1'-biphenyl]-2-carboxylic acid ≥97%
2',6'-dimethoxy-[1,1'-biphenyl]-2-carboxylic acid

Precaución: For research use only

Formula: C₁₅H₁₄O₄
MW: 258.269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11554156
CAS Number: 35394-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR