Methyl 7-bromo-4-chloro-3-(trifluoroMethyl)benzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

117011-1G 117011-500MG 117011-5G 117011-25G
77464-094EA 415.09 USD
77464-094 77476-886 77486-964 77497-820
Methyl 7-bromo-4-chloro-3-(trifluoroMethyl)benzo[b]thiophene-2-carboxylate ≥95%
Methyl 7-bromo-4-chloro-3-(trifluoroMethyl)benzo[b]thiophene-2-carboxylate

Precaución: For research use only

Formula: C₁₁H₅BrClF₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD34783295
CAS Number: 2918910-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR