2,4-Dichloro-3-(1,3-Dioxolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

116992-1G 116992-500MG 116992-5G 116992-25G
77464-090EA 415.09 USD
77464-090 77476-882 77486-960 77497-818
2,4-Dichloro-3-(1,3-Dioxolan-2-yl)pyridine ≥97%
2,4-Dichloro-3-(1,3-Dioxolan-2-yl)pyridine

Precaución: For research use only

Formula: C₈H₇Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD24368909
CAS Number: 1427080-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR