(4-Fluoro-2-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

116712-1G 116712-500MG 116712-5G 116712-25G
77464-086EA 415.09 USD
77464-086 77476-878 77486-956 77497-812
(4-Fluoro-2-methylphenyl)(phenyl)methanol ≥95%
(4-Fluoro-2-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₃FO
MW: 216.2508 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222655
CAS Number: 1340291-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR