(3-Bromo-2-chloro-6-fluorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

116699-1G 116699-500MG 116699-5G 116699-25G
77464-082EA 415.09 USD
77464-082 77476-874 77486-952 77497-808
(3-Bromo-2-chloro-6-fluorophenyl)(phenyl)methanol ≥95%
(3-Bromo-2-chloro-6-fluorophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₉BrClFO
MW: 315.5653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823270
CAS Number: 2969484-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR