1-Bromo-2-isopropoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

116665-1G 116665-500MG 116665-5G
77464-078EA 576.8 USD
77464-078 77476-870 77486-948
1-Bromo-2-isopropoxy-3-methylbenzene ≥95%
1-Bromo-2-isopropoxy-3-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD12547893
CAS Number: 1208076-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR