2-Ethoxy-1-iodo-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

116656-1G 116656-500MG 116656-5G 116656-25G
77464-074EA 415.09 USD
77464-074 77476-866 77486-944 77497-804
2-Ethoxy-1-iodo-3-methylbenzene ≥95%
2-Ethoxy-1-iodo-3-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁IO
MW: 262.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547888
CAS Number: 861349-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR