(2-Ethoxy-3-methylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

116650-1G 116650-500MG 116650-5G
77464-070EA 787.95 USD
77464-070 77476-862 77486-940
(2-Ethoxy-3-methylphenyl)boronic acid ≥95%
(2-Ethoxy-3-methylphenyl)boronic acid

Precaución: For research use only

Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD28384208
CAS Number: 1704064-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR