1,2,4-tribromo-3,5-dimethylbenzene ≥96%

Supplier: AOBChem USA

Warning

116491-1G 116491-500MG 116491-5G 116491-25G
77464-066EA 347.11 USD
77464-066 77476-858 77486-936 77497-798
1,2,4-tribromo-3,5-dimethylbenzene ≥96%
1,2,4-tribromo-3,5-dimethylbenzene

Precaución: For research use only

Formula: C₈H₇Br₃
MW: 342.853 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823194
CAS Number: 860748-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR