3-Hydroxy-2,4-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

116402-1G 116402-500MG 116402-5G 116402-25G
77464-062EA 396.55 USD
77464-062 77476-854 77486-932 77497-794
3-Hydroxy-2,4-dimethoxybenzaldehyde ≥97%
3-Hydroxy-2,4-dimethoxybenzaldehyde

Precaución: For research use only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021066
CAS Number: 32246-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR