1-(2-Methoxy-5-(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

116207-1G 116207-500MG 116207-5G
77464-058EA 447.02 USD
77464-058 77476-850 77486-928
1-(2-Methoxy-5-(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(2-Methoxy-5-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃F₃O₂
MW: 234.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22003415
CAS Number: 1472033-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR