Ethyl-3-chloro-4-hydroxybenzoate ≥95%

Supplier: AOBChem USA

Warning

115898-1G 115898-500MG 115898-5G 115898-25G
77464-054EA 74.16 USD
77464-054 77476-846 77486-924 77497-788
Ethyl-3-chloro-4-hydroxybenzoate ≥95%
Ethyl-3-chloro-4-hydroxybenzoate

Precaución: For research use only

Formula: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD05664241
CAS Number: 16357-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR