1-(2-Fluoro-5-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

115520-1G 115520-500MG 115520-5G 115520-25G
77464-050EA 415.09 USD
77464-050 77476-842 77486-920 77497-780
1-(2-Fluoro-5-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(2-Fluoro-5-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₄F₄O
MW: 250.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24930719
CAS Number: 1524210-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR