1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

115508-1G 115508-500MG 115508-5G 115508-25G
77464-046EA 415.09 USD
77464-046 77476-838 77486-916 77497-776
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Bromo-6-fluoro-3-methoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO₂
MW: 291.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34823105
CAS Number: 2969374-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR