1-(3-Bromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥96%

Supplier: AOBChem USA

Warning

115500-1G 115500-500MG 115500-5G
77464-042EA 447.02 USD
77464-042 77476-834 77486-912
1-(3-Bromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol ≥96%
1-(3-Bromo-2-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
Storage Temperature: Ambient
MDL Number: MFCD34823102
CAS Number: 1694516-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR