1-(2,3-Difluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

115495-1G 115495-500MG 115495-5G 115495-25G
77464-038EA 415.09 USD
77464-038 77476-830 77486-908 77497-772
1-(2,3-Difluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,3-Difluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄F₂O
MW: 200.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19626924
CAS Number: 1342480-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR