1-(3-Methoxyphenyl)-2-methylpropan-1-one ≥97%

Supplier: AOBChem USA

Warning

115467-1G 115467-500MG 115467-5G 115467-25G
77464-034EA 331.66 USD
77464-034 77476-826 77486-904 77497-768
1-(3-Methoxyphenyl)-2-methylpropan-1-one ≥97%
1-(3-Methoxyphenyl)-2-methylpropan-1-one

Precaución: For research use only

Formula: C₁₁H₁₄O₂
MW: 178.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153242
CAS Number: 6026-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR