1-(3-Bromophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

115460-1G 115460-500MG 115460-5G 115460-25G
77464-030EA 260.59 USD
77464-030 77476-822 77486-900 77497-764
1-(3-Bromophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Bromophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16748773
CAS Number: 92644-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR