1-(4-Methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

115452-1G 115452-500MG 115452-5G 115452-25G
77464-026EA 260.59 USD
77464-026 77476-818 77486-896 77497-762
1-(4-Methoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Methoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153740
CAS Number: 6013-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR