4'-Methoxy-4,4-dimethyl-2,3,4,5-tetrahydro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

115417-1G 115417-500MG 115417-5G 115417-25G
77464-022EA 415.09 USD
77464-022 77476-814 77486-892 77497-756
4'-Methoxy-4,4-dimethyl-2,3,4,5-tetrahydro-1,1'-biphenyl ≥95%
4'-Methoxy-4,4-dimethyl-2,3,4,5-tetrahydro-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₅H₂₀O
MW: 216.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34782695
CAS Number: 151772-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR