2-Chloro-3-iodo-5-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

11502-1G 11502-5G 11502-25G
77464-018EA 63.86 USD
77464-018 77486-886 77497-750
2-Chloro-3-iodo-5-(trifluoromethyl)pyridine ≥97%
2-Chloro-3-iodo-5-(trifluoromethyl)pyridine

Precaución: For research use only

Formula: C₆H₂ClF₃IN
Storage Temperature: Ambient
MDL Number: MFCD08741352
CAS Number: 505084-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR