2-[(3-Methylbenzyl)oxy]benzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
114245-1G
114245-500MG
114245-5G
114245-25G
77464-014EA
95.79
USD
77464-014
77476-810
77486-880
77497-738
2-[(3-Methylbenzyl)oxy]benzoic acid ≥95%
2-[(3-Methylbenzyl)oxy]benzoic acid
Precaución: For research use only
Formula:
C₁₅H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD05267697 CAS Number: 744242-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...