N-Benzyl-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

114007-1G 114007-500MG 114007-5G 114007-25G
77464-010EA 197.76 USD
77464-010 77476-806 77486-876 77497-732
N-Benzyl-4-methylbenzamide ≥97%
N-Benzyl-4-methylbenzamide

Precaución: For research use only

Formula: C₁₅H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD00026008
CAS Number: 5436-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR