2-Bromo-6-chloro-5-methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

113750-1G 113750-500MG 113750-5G 113750-25G
77464-006EA 415.09 USD
77464-006 77476-802 77486-872 77497-730
2-Bromo-6-chloro-5-methylpyridin-3-ol ≥97%
2-Bromo-6-chloro-5-methylpyridin-3-ol

Precaución: For research use only

Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26383851
CAS Number: 1823355-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR