4-(3-Chloro-2,6-difluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

113449-1G 113449-500MG 113449-5G 113449-25G
77464-002EA 415.09 USD
77464-002 77476-798 77486-868 77497-726
4-(3-Chloro-2,6-difluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3-Chloro-2,6-difluorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₁H₁₁ClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34781865
CAS Number: 2918905-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR