(4-(tert-Butyl)-2-isopropoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

112846-1G 112846-500MG 112846-5G 112846-25G
77463-998EA 286.34 USD
77463-998 77476-794 77486-860 77497-718
(4-(tert-Butyl)-2-isopropoxyphenyl)trimethylsilane ≥97%
(4-(tert-Butyl)-2-isopropoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₆H₂₈OSi
MW: 264.478 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34822440
CAS Number: 2969332-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR