1-(4-(tert-Butyl)-2-ethoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

112834-1G 112834-500MG 112834-5G 112834-25G
77463-994EA 415.09 USD
77463-994 77476-790 77486-856 77497-714
1-(4-(tert-Butyl)-2-ethoxyphenyl)ethanone ≥95%
1-(4-(tert-Butyl)-2-ethoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₄H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34822436
CAS Number: 2969462-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR