4-(tert-Butyl)-2-ethoxy-1-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

112832-1G 112832-5G 112832-25G
77463-990EA 60.77 USD
77463-990 77486-852 77497-710
4-(tert-Butyl)-2-ethoxy-1-iodobenzene ≥95%
4-(tert-Butyl)-2-ethoxy-1-iodobenzene

Precaución: For research use only

Formula: C₁₂H₁₇IO
Storage Temperature: Ambient
MDL Number: MFCD28991748
CAS Number: 870007-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR