(2,3-Difluoro-4-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

112448-1G 112448-500MG 112448-5G 112448-25G
77463-986EA 415.09 USD
77463-986 77476-784 77486-848 77497-704
(2,3-Difluoro-4-methylphenyl)(ethyl)sulfane ≥95%
(2,3-Difluoro-4-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀F₂S
Storage Temperature: Ambient
MDL Number: MFCD34781553
CAS Number: 2918882-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR