3,5-Dichloro-2-fluorobenzoyl chloride ≥95%

Supplier: AOBChem USA

Warning

111623-1G 111623-500MG 111623-5G
77463-982EA 676.71 USD
77463-982 77476-780 77486-844
3,5-Dichloro-2-fluorobenzoyl chloride ≥95%
3,5-Dichloro-2-fluorobenzoyl chloride

Precaución: For research use only

Formula: C₇H₂Cl₃FO
MW: 227.448 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687667
CAS Number: 1529604-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR