6-Bromo-7-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

111362-1G 111362-500MG 111362-5G 111362-25G
77463-978EA 131.84 USD
77463-978 77476-778 77486-840 77497-696
6-Bromo-7-methyl-1H-indazole ≥97%
6-Bromo-7-methyl-1H-indazole

Precaución: For research use only

Formula: C₈H₇BrN₂
MW: 211.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23705395
CAS Number: 1337880-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR