4-(2,5-Dichlorophenyl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

111219-1G 111219-500MG 111219-5G 111219-25G
77463-974EA 226.6 USD
77463-974 77476-774 77486-836 77497-690
4-(2,5-Dichlorophenyl)tetrahydro-2H-pyran-4-ol ≥97%
4-(2,5-Dichlorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₁H₁₂Cl₂O₂
MW: 247.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18296155
CAS Number: 1340313-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR