2-Bromo-6-fluorophenyl acetate ≥95%

Supplier: AOBChem USA

Warning

111199-1G 111199-500MG 111199-5G 111199-25G
77463-970EA 415.09 USD
77463-970 77476-770 77486-832 77497-686
2-Bromo-6-fluorophenyl acetate ≥95%
2-Bromo-6-fluorophenyl acetate

Precaución: For research use only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34780964
CAS Number: 2901103-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR